missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Fluorophenyl)ethanol 95.0+%, TCI America™

Catalog No. F07785G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07785G 5 g
F077825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07785G Supplier TCI America Supplier No. F07785G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50919-06-7
Molecular Formula C8H9FO
Molecular Weight (g/mol) 140.16
MDL Number MFCD00002887
InChI Key HNIGZVZDWCTFPR-UHFFFAOYSA-N
Synonym 2-fluorophenethyl alcohol, 2-2-fluorophenyl ethanol, 2-2-fluorophenyl ethan-1-ol, benzeneethanol, 2-fluoro, 2-fluorophenethylalcohol, 2-2-fluoro-phenyl-ethanol, 2-fluorobenzeneethanol, acmc-1axdv, o-fluorophenethyl alcohol, 2-fluorophenylethyl alcohol
PubChem CID 2733296
IUPAC Name 2-(2-fluorophenyl)ethan-1-ol
SMILES OCCC1=CC=CC=C1F

Specifications

Color Yellow
Boiling Point 137°C
Quantity 5 g
Formula Weight 140.16
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(2-Fluorophenyl)ethanol