missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Fluorophenyl)ethanol, 99%

Catalog No. p-7058529
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
AAL1999806 5 g
AAL1999814 25 g
2 options

Catalog No. AAL1999806

Supplier: Thermo Scientific Chemicals L1999806

Only null left

CAS: 50919-06-7 | C8H9FO | 140.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50919-06-7
Molecular Formula C8H9FO
Molecular Weight (g/mol) 140.16
MDL Number MFCD00002887
InChI Key HNIGZVZDWCTFPR-UHFFFAOYSA-N
Synonym 2-fluorophenethyl alcohol, 2-2-fluorophenyl ethanol, 2-2-fluorophenyl ethan-1-ol, benzeneethanol, 2-fluoro, 2-fluorophenethylalcohol, 2-2-fluoro-phenyl-ethanol, 2-fluorobenzeneethanol, acmc-1axdv, o-fluorophenethyl alcohol, 2-fluorophenylethyl alcohol
PubChem CID 2733296
IUPAC Name 2-(2-fluorophenyl)ethanol
SMILES OCCC1=CC=CC=C1F

Specifications

Density 1.045
Boiling Point 72°C (1 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.51
Quantity 5 g
Formula Weight 140.16
Percent Purity 99%
Chemical Name or Material 2-(2-Fluorophenyl)ethanol
Hazard Category H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only