missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Ethylenedioxydiphenol 98.0+%, TCI America™

Catalog No. E05571G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E05571G Supplier TCI America Supplier No. E05571G
Only null left

Chemical Identifiers

CAS 20115-81-5
Molecular Formula C14H14O4
Molecular Weight (g/mol) 246.262
MDL Number MFCD00441177
InChI Key PRYNAEBLJBDWPF-UHFFFAOYSA-N
Synonym 1,2-Bis(2-hydroxyphenoxy)ethane, Ethylene Glycol Bis(2-hydroxyphenyl) Ether
PubChem CID 5171332
IUPAC Name 2-[2-(2-hydroxyphenoxy)ethoxy]phenol
SMILES C1=CC=C(C(=C1)O)OCCOC2=CC=CC=C2O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 246.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Ethylenedioxydiphenol