accessibility menu, dialog, popup

UserName

OHOOHO

CAS RN 20115-81-5

IUPAC Name: 2-[2-(2-hydroxyphenoxy)ethoxy]phenol
Synonyms: 2-[2-(2-hydroxyphenoxy)ethoxy]phenol
Molecular Weight (g/mol): 246.26
Molecular Formula: C14H14O4
InChi Key: PRYNAEBLJBDWPF-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1OCCOC1=CC=CC=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results