missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Dipyridyl Disulfide 98.0+%, TCI America™
Supplier: TCI America D1114250G
Specifications
| 2,2′-Dipyridyl Disulfide [for Peptide Synthesis] | |
| 58°C | |
| C10H8N2S2 | |
| 250 g | |
| HAXFWIACAGNFHA-UHFFFAOYSA-N | |
| 2-(pyridin-2-yldisulfanyl)pyridine | |
| 65093 | |
| ≥98.0% (GC,T) |
| 2127-03-9 | |
| White | |
| MFCD00006287 | |
| 2,2'-dithiodipyridine, 2,2'-dipyridyl disulfide, 1,2-di pyridin-2-yl disulfane, aldrithiol-2, 2,2'-dipyridyldisulfide, aldrithiol 2, bis 2-pyridyl disulfide, 2-pyridin-2-yldisulfanyl pyridine, 2-dipyridyl disulfide, 2-pyridinyl disulfide | |
| C1=CC=NC(=C1)SSC2=CC=CC=N2 | |
| 220.31 | |
| 220.31 | |
| Crystalline Powder |
Chemical Identifiers
| 2127-03-9 | |
| 220.31 | |
| HAXFWIACAGNFHA-UHFFFAOYSA-N | |
| 65093 | |
| C1=CC=NC(=C1)SSC2=CC=CC=N2 |
| C10H8N2S2 | |
| MFCD00006287 | |
| 2,2'-dithiodipyridine, 2,2'-dipyridyl disulfide, 1,2-di pyridin-2-yl disulfane, aldrithiol-2, 2,2'-dipyridyldisulfide, aldrithiol 2, bis 2-pyridyl disulfide, 2-pyridin-2-yldisulfanyl pyridine, 2-dipyridyl disulfide, 2-pyridinyl disulfide | |
| 2-(pyridin-2-yldisulfanyl)pyridine |
Safety and Handling
EINECSNumber : (5)-0730
TSCA : Yes