missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dipyridyl Disulfide 98.0+%, TCI America™

Catalog No. p-7126603
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D11145G 5 g
D111425G 25 g
D1114250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D11145G Supplier TCI America Supplier No. D11145G
Only null left

Chemical Identifiers

CAS 2127-03-9
Molecular Formula C10H8N2S2
Molecular Weight (g/mol) 220.31
MDL Number MFCD00006287
InChI Key HAXFWIACAGNFHA-UHFFFAOYSA-N
Synonym 2,2'-dithiodipyridine, 2,2'-dipyridyl disulfide, 1,2-di pyridin-2-yl disulfane, aldrithiol-2, 2,2'-dipyridyldisulfide, aldrithiol 2, bis 2-pyridyl disulfide, 2-pyridin-2-yldisulfanyl pyridine, 2-dipyridyl disulfide, 2-pyridinyl disulfide
PubChem CID 65093
IUPAC Name 2-(pyridin-2-yldisulfanyl)pyridine
SMILES C1=CC=NC(=C1)SSC2=CC=CC=N2

Specifications

Melting Point 58°C
Color White
Quantity 5 g
Formula Weight 220.31
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Dipyridyl Disulfide [for Peptide Synthesis]
EINECSNumber (5)-0730
TSCA Yes