missing translation for 'onlineSavingsMsg'
Learn More

2,2-Diphenylpropionic Acid 96.0+%, TCI America™

Catalog No. D17785G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D17785G Supplier TCI America Supplier No. D17785G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5558-66-7
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.275
MDL Number MFCD00004189
InChI Key ODELFXJUOVNEFZ-UHFFFAOYSA-N
Synonym 2,2-diphenylpropionic acid, 2,2-diphenylpropionicacid, a,a-diphenylpropionic acid, alpha,alpha-diphenylpropionic acid, acmc-209lnv, cambridge id 7170182, 2,2-diphenyl propanoic acid, 2,2-diphenyl propionic acid, 2,2-di phenyl propanoic acid, #
PubChem CID 79676
IUPAC Name 2,2-diphenylpropanoic acid
SMILES CC(C1=CC=CC=C1)(C2=CC=CC=C2)C(=O)O

Specifications

Melting Point 175°C
Color White-Yellow
Quantity 5 g
Formula Weight 226.28
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Diphenylpropionic Acid