missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dinitrobiphenyl 99.0+%, TCI America™

Supplier:  TCI America D177725G

 View more versions of this product

Catalog No. D177725G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,2′-Dinitrobiphenyl
125°C
MFCD00007127
2,2'-dinitrobiphenyl, 2,2'-dinitro-1,1'-biphenyl, 1,1'-biphenyl, 2,2'-dinitro, 2,2'-dinitro-biphenyl, 1-nitro-2-2-nitrophenyl benzene, 2,2'-dinitrodiphenyl, biphenyl, 2,2'-dinitro, o,o'-binitrobiphenyl, o,o'-dinitrobiphenyl, 2,2'-dinitrobiphenyl, 2,2'-dinitro
C1=CC=C(C(=C1)C2=CC=CC=C2[N+](=O)[O-])[N+](=O)[O-]
244.206
CHEBI:67127
≥99.0% (GC)
2436-96-6
C12H8N2O4
25 g
QAFJHDNFUMKVIE-UHFFFAOYSA-N
1-nitro-2-(2-nitrophenyl)benzene
75529
244.21
Crystalline Powder

Chemical Identifiers

2436-96-6
244.206
QAFJHDNFUMKVIE-UHFFFAOYSA-N
75529
1-nitro-2-(2-nitrophenyl)benzene
C12H8N2O4
MFCD00007127
2,2'-dinitrobiphenyl, 2,2'-dinitro-1,1'-biphenyl, 1,1'-biphenyl, 2,2'-dinitro, 2,2'-dinitro-biphenyl, 1-nitro-2-2-nitrophenyl benzene, 2,2'-dinitrodiphenyl, biphenyl, 2,2'-dinitro, o,o'-binitrobiphenyl, o,o'-dinitrobiphenyl, 2,2'-dinitrobiphenyl, 2,2'-dinitro
CHEBI:67127
C1=CC=C(C(=C1)C2=CC=CC=C2[N+](=O)[O-])[N+](=O)[O-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions