missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dinitrobiphenyl 99.0+%, TCI America™

Catalog No. p-7126921
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17775G 5 g
D177725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D17775G Supplier TCI America Supplier No. D17775G

Chemical Identifiers

CAS 2436-96-6
Molecular Formula C12H8N2O4
Molecular Weight (g/mol) 244.206
MDL Number MFCD00007127
InChI Key QAFJHDNFUMKVIE-UHFFFAOYSA-N
Synonym 2,2'-dinitrobiphenyl, 2,2'-dinitro-1,1'-biphenyl, 1,1'-biphenyl, 2,2'-dinitro, 2,2'-dinitro-biphenyl, 1-nitro-2-2-nitrophenyl benzene, 2,2'-dinitrodiphenyl, biphenyl, 2,2'-dinitro, o,o'-binitrobiphenyl, o,o'-dinitrobiphenyl, 2,2'-dinitrobiphenyl, 2,2'-dinitro
PubChem CID 75529
ChEBI CHEBI:67127
IUPAC Name 1-nitro-2-(2-nitrophenyl)benzene
SMILES C1=CC=C(C(=C1)C2=CC=CC=C2[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 125°C
Quantity 5 g
Formula Weight 244.21
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Dinitrobiphenyl