missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylthiazolidine 98.0+%, TCI America™

Catalog No. p-7126747
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D14915ML 5 mL
D149125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D14915ML Supplier TCI America Supplier No. D14915ML
Only null left

Chemical Identifiers

CAS 19351-18-9
Molecular Formula C5H12ClNS
Molecular Weight (g/mol) 153.67
MDL Number MFCD00014488
InChI Key NKPNSVCEUIECCA-UHFFFAOYSA-N
PubChem CID 88015
IUPAC Name hydrogen 2,2-dimethyl-1,3-thiazolidine chloride
SMILES [H+].[Cl-].CC1(C)NCCS1

Specifications

Color Yellow
Boiling Point 61°C
Quantity 5 mL
UN Number 1993
Formula Weight 117.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2-Dimethylthiazolidine
EINECSNumber (5)-5890
RTECSNumber XJ5565700
TSCA Yes