accessibility menu, dialog, popup

UserName

CAS RN 19351-18-9

IUPAC Name: hydrogen 2,2-dimethyl-1,3-thiazolidine chloride
Synonyms: hydrogen 2,2-dimethylthiazolidine chloride
Molecular Weight (g/mol): 153.67
Molecular Formula: C5H12ClNS
InChi Key: NKPNSVCEUIECCA-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1(C)NCCS1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results