missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Dihydroxybenzophenone 99.0+%, TCI America™
$374.68 - $747.49
Chemical Identifiers
| CAS | 835-11-0 |
|---|---|
| Molecular Formula | C13H10O3 |
| Molecular Weight (g/mol) | 214.22 |
| MDL Number | MFCD00002217 |
| InChI Key | YIYBRXKMQFDHSM-UHFFFAOYSA-N |
| PubChem CID | 70038 |
| ChEBI | CHEBI:34209 |
| IUPAC Name | bis(2-hydroxyphenyl)methanone |
| SMILES | C1=CC=C(C(=C1)C(=O)C2=CC=CC=C2O)O |
Chemical Identifiers
| 835-11-0 | |
| 214.22 | |
| YIYBRXKMQFDHSM-UHFFFAOYSA-N | |
| CHEBI:34209 | |
| C1=CC=C(C(=C1)C(=O)C2=CC=CC=C2O)O |
| C13H10O3 | |
| MFCD00002217 | |
| 70038 | |
| bis(2-hydroxyphenyl)methanone |
Specifications
| 835-11-0 | |
| Yellow | |
| MFCD00002217 | |
| YIYBRXKMQFDHSM-UHFFFAOYSA-N | |
| bis(2-hydroxyphenyl)methanone | |
| 70038 | |
| 214.22 | |
| Crystalline Powder |
| 63°C | |
| C13H10O3 | |
| 10 g | |
| C1=CC=C(C(=C1)C(=O)C2=CC=CC=C2O)O | |
| 214.22 | |
| CHEBI:34209 | |
| ≥99.0% (GC) | |
| 2,2′-Dihydroxybenzophenone |
Safety and Handling
TSCA : No