missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Dihydroxybenzophenone 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 63°C |
| Color | Yellow |
| Quantity | 10 g |
| Formula Weight | 214.22 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,2′-Dihydroxybenzophenone |
Safety and Handling
| TSCA | No |