missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dibromopropane 98.0+%, TCI America™

Catalog No. D140910G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D140910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D140910G Supplier TCI America Supplier No. D140910G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 594-16-1
Molecular Formula C3H6Br2
Molecular Weight (g/mol) 201.89
MDL Number MFCD00000119
InChI Key ARITXYXYCOZKMU-UHFFFAOYSA-N
Synonym propane, 2,2-dibromo, 2,2-dibromopropane, dibromo propane, 2-dibrompropan, acmc-1aolk, ksc272i9f, inchi=1/c3h6br2/c1-3 2,4 5/h1-2h
PubChem CID 11658
IUPAC Name 2,2-dibromopropane
SMILES CC(C)(Br)Br

Specifications

Boiling Point 113°C
Quantity 10 g
Formula Weight 201.89
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2-Dibromopropane