accessibility menu, dialog, popup

UserName

CAS RN 594-16-1

IUPAC Name: 2,2-dibromopropane
Synonyms: propane, 2,2-dibromo-
Molecular Weight (g/mol): 201.89
Molecular Formula: C3H6Br2
InChi Key: ARITXYXYCOZKMU-UHFFFAOYSA-N
SMILES: CC(C)(Br)Br
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)