missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dibromopropane, 98%

Catalog No. p-102597
Change view
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC186060050 5 g Glass bottle
AC186060500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC186060050 Supplier Thermo Scientific Chemicals Supplier No. 186060050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 594-16-1 | C3H6Br2 | 201.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 594-16-1
Molecular Formula C3H6Br2
Molecular Weight (g/mol) 201.89
MDL Number MFCD00000119
InChI Key ARITXYXYCOZKMU-UHFFFAOYSA-N
Synonym propane, 2,2-dibromo, 2,2-dibromopropane, dibromo propane, 2-dibrompropan, acmc-1aolk, ksc272i9f, inchi=1/c3h6br2/c1-3 2,4 5/h1-2h
PubChem CID 11658
IUPAC Name 2,2-dibromopropane
SMILES CC(C)(Br)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.7800g/mL
Boiling Point 114°C (740.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4950 to 1.499
Linear Formula CH3CBr2CH3
Quantity 5 g
Beilstein 01, 111
Specific Gravity 1.78
Formula Weight 201.89
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 2-Dibromopropane, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-828-9
TSCA TSCA

RUO – Research Use Only