missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dibenzothiazolyl Disulfide 96.0+%, TCI America™

Catalog No. p-7126276
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D053825G 25 g
D0538500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D053825G Supplier TCI America Supplier No. D053825G
Only null left

Chemical Identifiers

CAS 120-78-5
Molecular Formula C14H8N2S4
Molecular Weight (g/mol) 332.472
MDL Number MFCD00022874
InChI Key AFZSMODLJJCVPP-UHFFFAOYSA-N
Synonym altax, thiofide, dibenzothiazyl disulfide, benzothiazyl disulfide, 2,2'-dithiobis benzothiazole, 2,2'-dithiobisbenzothiazole, benzothiazole disulfide, mbts, pneumax dm, vulcafor mbts
PubChem CID 8447
ChEBI CHEBI:53239
IUPAC Name 2-(1,3-benzothiazol-2-yldisulfanyl)-1,3-benzothiazole
SMILES C1=CC=C2C(=C1)N=C(S2)SSC3=NC4=CC=CC=C4S3

Specifications

Melting Point 178°C
Quantity 25 g
UN Number 3077
Formula Weight 332.47
Percent Purity ≥96.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Dibenzothiazolyl Disulfide
EINECSNumber (5)-0265
RTECSNumber DL4550000
TSCA Yes