accessibility menu, dialog, popup

UserName

CAS RN 120-78-5

IUPAC Name: 2-(1,3-benzothiazol-2-yldisulfanyl)-1,3-benzothiazole
Synonyms: altax MBTS 2,2'-benzothiazyl disulfide thiofide 2,2'-dibenzothiazyl disulfide bts-sbt
Molecular Weight (g/mol): 332.47
Molecular Formula: C14H8N2S4
InChi Key: AFZSMODLJJCVPP-UHFFFAOYSA-N
SMILES: S(SC1=NC2=CC=CC=C2S1)C1=NC2=CC=CC=C2S1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results