missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Diaminobiphenyl 99.0+%, TCI America™
Supplier: TCI America D52555G
Specifications
| 2,2′-Diaminobiphenyl | |
| 78°C | |
| C12H12N2 | |
| 5 g | |
| HOLGXWDGCVTMTB-UHFFFAOYSA-N | |
| 2-(2-aminophenyl)aniline | |
| 15075 | |
| ≥99.0% (GC,T) |
| 1454-80-4 | |
| 151°C | |
| MFCD00034060 | |
| [1,1′-Biphenyl]-2,2′-diamine | |
| C1=CC=C(C(=C1)C2=CC=CC=C2N)N | |
| 184.242 | |
| 184.24 | |
| Crystalline Powder |
Chemical Identifiers
| 1454-80-4 | |
| 184.242 | |
| HOLGXWDGCVTMTB-UHFFFAOYSA-N | |
| 15075 | |
| C1=CC=C(C(=C1)C2=CC=CC=C2N)N |
| C12H12N2 | |
| MFCD00034060 | |
| [1,1′-Biphenyl]-2,2′-diamine | |
| 2-(2-aminophenyl)aniline |
Safety and Handling
RTECSNumber : DV2099000
TSCA : No