missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Diaminobiphenyl 99.0+%, TCI America™

Catalog No. p-7128709
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
D52551G 1 g
D52555G 5 g
2 options

Catalog No. D52551G

Supplier: TCI America D52551G

Only null left

Chemical Identifiers

CAS 1454-80-4
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.242
MDL Number MFCD00034060
InChI Key HOLGXWDGCVTMTB-UHFFFAOYSA-N
Synonym [1,1′-Biphenyl]-2,2′-diamine
PubChem CID 15075
IUPAC Name 2-(2-aminophenyl)aniline
SMILES C1=CC=C(C(=C1)C2=CC=CC=C2N)N

Specifications

Melting Point 78°C
Boiling Point 151°C
Quantity 1 g
Formula Weight 184.24
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Diaminobiphenyl
RTECSNumber DV2099000
TSCA No