accessibility menu, dialog, popup

UserName

NH2NH2

CAS RN 1454-80-4

IUPAC Name: [1,1'-biphenyl]-2,2'-diamine
Synonyms: [1,1'-biphenyl]-2,2'-diamine
Molecular Weight (g/mol): 184.24
Molecular Formula: C12H12N2
InChi Key: HOLGXWDGCVTMTB-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1C1=CC=CC=C1N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results