missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97.0+%, TCI America™

Catalog No. B28281G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28281G 1 g
B28285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28281G Supplier TCI America Supplier No. B28281G
Only null left

Chemical Identifiers

CAS 269410-06-2
Molecular Formula C12H16BBrO2
Molecular Weight (g/mol) 282.972
InChI Key BQVWGVYJHSRHSD-UHFFFAOYSA-N
Synonym 1-Bromo-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 2-Bromophenylboronic Acid Pinacol Ester
PubChem CID 21923944
IUPAC Name 2-(2-bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2Br

Specifications

Color Yellow
Quantity 1 g
Formula Weight 282.97
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 2-(2-Bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane