CAS RN 269410-06-2
IUPAC Name:
2-(2-bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms:
2-(2-bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol):
282.97
Molecular Formula:
C12H16BBrO2
InChi Key:
BQVWGVYJHSRHSD-UHFFFAOYSA-N
SMILES:
CC1(C)OB(OC1(C)C)C1=CC=CC=C1Br
1
–
1
of
1
results
1
2-(2-Bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97.0+%, TCI America™
CAS: 269410-06-2 Molecular Formula: C12H16BBrO2 Molecular Weight (g/mol): 282.972 InChI Key: BQVWGVYJHSRHSD-UHFFFAOYSA-N Synonym: 1-Bromo-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 2-Bromophenylboronic Acid Pinacol Ester PubChem CID: 21923944 IUPAC Name: 2-(2-bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane SMILES: B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2Br