missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Bis(hydroxymethyl)diphenyl Ether 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 99°C |
| Color | White |
| Quantity | 1 g |
| Synonym | 2,2′C-Oxydi(benzyl Alcohol) |
| Formula Weight | 230.26 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,2′-Bis(hydroxymethyl)diphenyl Ether |
Safety and Handling
| TSCA | No |