accessibility menu, dialog, popup

UserName

OHOOH

CAS RN 10038-40-1

IUPAC Name: {2-[2-(hydroxymethyl)phenoxy]phenyl}methanol
Synonyms: {2-[2-(hydroxymethyl)phenoxy]phenyl}methanol
Molecular Weight (g/mol): 230.26
Molecular Formula: C14H14O3
InChi Key: VRVKKKKXKVCPEW-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=C1OC1=CC=CC=C1CO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results