missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Bis(4-isocyanatophenyl)hexafluoropropane 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 1 g |
| UN Number | 2206 |
| Formula Weight | 386.25 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2,2-Bis(4-isocyanatophenyl)hexafluoropropane |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |