CAS RN 10224-18-7
IUPAC Name:
1-[1,1,1,3,3,3-hexafluoro-2-(4-isocyanatophenyl)propan-2-yl]-4-isocyanatobenzene
Synonyms:
1-[1,1,1,3,3,3-hexafluoro-2-(4-isocyanatophenyl)propan-2-yl]-4-isocyanatobenzene
Molecular Weight (g/mol):
386.25
Molecular Formula:
C17H8F6N2O2
InChi Key:
QIPLQPPNURSGKC-UHFFFAOYSA-N
SMILES:
FC(F)(F)C(C1=CC=C(C=C1)N=C=O)(C1=CC=C(C=C1)N=C=O)C(F)(F)F
1
–
1
of
1
results
1
2,2-Bis(4-isocyanatophenyl)hexafluoropropane 98.0+%, TCI America™
CAS: 10224-18-7 Molecular Formula: C17H8F6N2O2 Molecular Weight (g/mol): 386.25 MDL Number: MFCD00142759 InChI Key: QIPLQPPNURSGKC-UHFFFAOYSA-N Synonym: 4,4′C-(Hexafluoroisopropylidene)bis(isocyanatobenzene) PubChem CID: 2736044 IUPAC Name: 1-[1,1,1,3,3,3-hexafluoro-2-(4-isocyanatophenyl)propan-2-yl]-4-isocyanatobenzene SMILES: FC(F)(F)C(C1=CC=C(C=C1)N=C=O)(C1=CC=C(C=C1)N=C=O)C(F)(F)F