missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(4-glycidyloxyphenyl)propane 85.0+%, TCI America™

Catalog No. p-7119268
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
B179625G 25 g
B1796500G 500 g
2 options

Catalog No. B179625G

Supplier: TCI America B179625G

Only null left

Chemical Identifiers

CAS 1675-54-3
Molecular Formula C21H24O4
Molecular Weight (g/mol) 340.419
MDL Number MFCD00080480
InChI Key LCFVJGUPQDGYKZ-UHFFFAOYSA-N
Synonym bisphenol a diglycidyl ether, 2,2-bis 4-glycidyloxyphenyl propane, epoxide a, epophen el 5, badge, dian diglycidyl ether, diglycidyl bisphenol a, epi-rez 510, bpdge, dgeba
PubChem CID 2286
ChEBI CHEBI:34578
IUPAC Name 2-[[4-[2-[4-(oxiran-2-ylmethoxy)phenyl]propan-2-yl]phenoxy]methyl]oxirane
SMILES CC(C)(C1=CC=C(C=C1)OCC2CO2)C3=CC=C(C=C3)OCC4CO4

Specifications

Color Yellow
Boiling Point 210°C
Quantity 25 g
Formula Weight 340.42
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2-Bis(4-glycidyloxyphenyl)propane
EINECSNumber (4)-0209
RTECSNumber TX3800000
TSCA Yes