accessibility menu, dialog, popup

UserName

CH3H3COOOO

CAS RN 1675-54-3

IUPAC Name: 2-{[4-(2-{4-[(oxiran-2-yl)methoxy]phenyl}propan-2-yl)phenoxy]methyl}oxirane
Synonyms: bisphenol A diglycidyl ether diglycidyl diphenylolpropane ether epi-rez 510 epi-rez 508 diglycidyl ether of bisphenol A dian diglycidyl ether bpdge dian-bis-glycidylether epophen EL 5 diglycidyl bisphenol A
Molecular Weight (g/mol): 340.42
Molecular Formula: C21H24O4
InChi Key: LCFVJGUPQDGYKZ-UHFFFAOYNA-N
SMILES: CC(C)(C1=CC=C(OCC2CO2)C=C1)C1=CC=C(OCC2CO2)C=C1
Quantity 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results