missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Bis(4-chlorophenyl)-1,1-dichloroethylene 99.0+%, TCI America™
$190.13 - $758.67
Chemical Identifiers
| CAS | 72-55-9 |
|---|---|
| Molecular Formula | C14H8Cl4 |
| Molecular Weight (g/mol) | 318.02 |
| MDL Number | MFCD00000837 |
| InChI Key | UCNVFOCBFJOQAL-UHFFFAOYSA-N |
| Synonym | p,p'-dde, dichlorodiphenyldichloroethylene, ddt dehydrochloride, 4,4'-dde, 2,2-bis 4-chlorophenyl-1,1-dichloroethylene, dde, 1,1-dichloro-2,2-bis 4-chlorophenyl ethene, p,p'-dichlorodiphenyldichloroethylene, p,p'-dichlorodiphenyl dichloroethylene, p,p'-dichlorodiphenyl-2,2-dichloroethylene |
| PubChem CID | 3035 |
| ChEBI | CHEBI:16598 |
| IUPAC Name | 1-chloro-4-[2,2-dichloro-1-(4-chlorophenyl)ethenyl]benzene |
| SMILES | ClC(Cl)=C(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=C1 |
Chemical Identifiers
| 72-55-9 | |
| 318.02 | |
| UCNVFOCBFJOQAL-UHFFFAOYSA-N | |
| 3035 | |
| 1-chloro-4-[2,2-dichloro-1-(4-chlorophenyl)ethenyl]benzene |
| C14H8Cl4 | |
| MFCD00000837 | |
| p,p'-dde, dichlorodiphenyldichloroethylene, ddt dehydrochloride, 4,4'-dde, 2,2-bis 4-chlorophenyl-1,1-dichloroethylene, dde, 1,1-dichloro-2,2-bis 4-chlorophenyl ethene, p,p'-dichlorodiphenyldichloroethylene, p,p'-dichlorodiphenyl dichloroethylene, p,p'-dichlorodiphenyl-2,2-dichloroethylene | |
| CHEBI:16598 | |
| ClC(Cl)=C(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=C1 |
Specifications
| 72-55-9 | |
| White | |
| MFCD00000837 | |
| 3077 | |
| UCNVFOCBFJOQAL-UHFFFAOYSA-N | |
| 1-chloro-4-[2,2-dichloro-1-(4-chlorophenyl)ethenyl]benzene | |
| 3035 | |
| 318.02 | |
| Crystalline Powder |
| 89°C | |
| C14H8Cl4 | |
| 1 g | |
| p,p'-dde, dichlorodiphenyldichloroethylene, ddt dehydrochloride, 4,4'-dde, 2,2-bis 4-chlorophenyl-1,1-dichloroethylene, dde, 1,1-dichloro-2,2-bis 4-chlorophenyl ethene, p,p'-dichlorodiphenyldichloroethylene, p,p'-dichlorodiphenyl dichloroethylene, p,p'-dichlorodiphenyl-2,2-dichloroethylene | |
| ClC(Cl)=C(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=C1 | |
| 318.02 | |
| CHEBI:16598 | |
| ≥99.0% (GC) | |
| 2,2-Bis(4-chlorophenyl)-1,1-dichloroethylene |
Safety and Handling
RTECSNumber : KV9450000
TSCA : No