accessibility menu, dialog, popup

UserName

ClClClCl

CAS RN 72-55-9

IUPAC Name: 1-chloro-4-[2,2-dichloro-1-(4-chlorophenyl)ethenyl]benzene
Synonyms: p,p'-DDE dichloroethylene, dichlorodiphenyl dichlorodiphenyldichloroethylene DDE DDX dichlorodiphenyl dichloroethene ddt dehydrochloride dichlorodiphenyl dichloroethylene ccris 193 P,P'-dde
Molecular Weight (g/mol): 318.02
Molecular Formula: C14H8Cl4
InChi Key: UCNVFOCBFJOQAL-UHFFFAOYSA-N
SMILES: ClC(Cl)=C(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results