missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(4-chlorophenyl)-1,1-dichloroethylene 99.0+%, TCI America™

Catalog No. B01331G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B01331G 1 g
B01335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B01331G Supplier TCI America Supplier No. B01331G
Only null left

Chemical Identifiers

CAS 72-55-9
Molecular Formula C14H8Cl4
Molecular Weight (g/mol) 318.02
MDL Number MFCD00000837
InChI Key UCNVFOCBFJOQAL-UHFFFAOYSA-N
Synonym p,p'-dde, dichlorodiphenyldichloroethylene, ddt dehydrochloride, 4,4'-dde, 2,2-bis 4-chlorophenyl-1,1-dichloroethylene, dde, 1,1-dichloro-2,2-bis 4-chlorophenyl ethene, p,p'-dichlorodiphenyldichloroethylene, p,p'-dichlorodiphenyl dichloroethylene, p,p'-dichlorodiphenyl-2,2-dichloroethylene
PubChem CID 3035
ChEBI CHEBI:16598
IUPAC Name 1-chloro-4-[2,2-dichloro-1-(4-chlorophenyl)ethenyl]benzene
SMILES ClC(Cl)=C(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=C1

Specifications

Melting Point 89°C
Color White
Quantity 1 g
UN Number 3077
Formula Weight 318.02
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis(4-chlorophenyl)-1,1-dichloroethylene
RTECSNumber KV9450000
TSCA No