missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(4-chlorophenyl)-1,1-dichloroethane 98.0+%, TCI America™

Catalog No. B01321G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B01321G 1 g
B01325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B01321G Supplier TCI America Supplier No. B01321G

Chemical Identifiers

CAS 72-54-8
Molecular Formula C14H10Cl4
Molecular Weight (g/mol) 320.034
MDL Number MFCD00000851
InChI Key AHJKRLASYNVKDZ-UHFFFAOYSA-N
Synonym p,p'-ddd, rhothane, dilene, tetrachlorodiphenylethane, dichlorodiphenyldichloroethane, p,p'-tde, rothane, rhothane d-3, 4,4'-ddd, 1,1-dichloro-2,2-bis p-chlorophenyl ethane
PubChem CID 6294
ChEBI CHEBI:27841
IUPAC Name 1-chloro-4-[2,2-dichloro-1-(4-chlorophenyl)ethyl]benzene
SMILES C1=CC(=CC=C1C(C2=CC=C(C=C2)Cl)C(Cl)Cl)Cl

Specifications

Melting Point 111°C
Color White
Quantity 1 g
UN Number 2811
Formula Weight 320.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis(4-chlorophenyl)-1,1-dichloroethane
RTECSNumber KI0700000
TSCA No