accessibility menu, dialog, popup

UserName

CAS RN 72-54-8

IUPAC Name: 1-chloro-4-[2,2-dichloro-1-(4-chlorophenyl)ethyl]benzene
Synonyms: dichlorodiphenyldichloroethane caswell no. 307 epa pesticide chemical code 029101 DDD rhothane D-3 TDE dilene ccris 573 rcra waste number U060 tetrachlorodiphenylethane
Molecular Weight (g/mol): 320.03
Molecular Formula: C14H10Cl4
InChi Key: AHJKRLASYNVKDZ-UHFFFAOYSA-N
SMILES: ClC(Cl)C(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results