missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(4-acetoxyphenyl)propane 98.0+%, TCI America™

Catalog No. p-7118971
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B128625G 25 g
B1286500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B128625G Supplier TCI America Supplier No. B128625G
Only null left

Chemical Identifiers

CAS 10192-62-8
Molecular Formula C19H20O4
Molecular Weight (g/mol) 312.37
MDL Number MFCD00026194
InChI Key NSNHONPMCQYMNT-UHFFFAOYSA-N
Synonym bisphenol a diacetate, 2,2-bis 4-acetoxyphenyl propane, phenol, 4,4'-1-methylethylidene bis-, diacetate, propane-2,2-diylbis 4,1-phenylene diacetate, phenol, 4,4'-1-methylethylidene bis-, 1,1'-diacetate, 4-2-4-acetyloxyphenyl propan-2-yl phenyl acetate, 4-2-4-acetyloxy phenyl propan-2-yl phenyl acetate, 4-1-4-acetyloxyphenyl-isopropyl phenyl acetate, 4,4'-isopropylidenediphenyl diacetate, propane-2,2-diyldi-4,1-phenylene diacetate
PubChem CID 82437
IUPAC Name 4-{2-[4-(acetyloxy)phenyl]propan-2-yl}phenyl acetate
SMILES CC(=O)OC1=CC=C(C=C1)C(C)(C)C1=CC=C(OC(C)=O)C=C1

Specifications

Melting Point 92°C
Color White
Boiling Point 215°C
Quantity 25 g
Formula Weight 312.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis(4-acetoxyphenyl)propane
EINECSNumber (4)-1661
TSCA Yes