accessibility menu, dialog, popup

UserName

CH3OOH3CCH3OCH3O

CAS RN 10192-62-8

IUPAC Name: 4-{2-[4-(acetyloxy)phenyl]propan-2-yl}phenyl acetate
Synonyms: 4-{2-[4-(acetyloxy)phenyl]propan-2-yl}phenyl acetate
Molecular Weight (g/mol): 312.37
Molecular Formula: C19H20O4
InChi Key: NSNHONPMCQYMNT-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=C(C=C1)C(C)(C)C1=CC=C(OC(C)=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results