missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis[4-(4-maleimidophenoxy)phenyl]propane 98.0+%, TCI America™

Catalog No. b480725g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
B480725G 25 g
B4807500G 500 g
2 options

Catalog No. B480725G

Supplier: TCI America B480725G

Only null left

Chemical Identifiers

CAS 79922-55-7
Molecular Formula C35H26N2O6
Molecular Weight (g/mol) 570.601
InChI Key XAZPKEBWNIUCKF-UHFFFAOYSA-N
Synonym Bisphenol A Bis(4-maleimidophenyl Ether)
PubChem CID 3086113
IUPAC Name 1-[4-[4-[2-[4-[4-(2,5-dioxopyrrol-1-yl)phenoxy]phenyl]propan-2-yl]phenoxy]phenyl]pyrrole-2,5-dione
SMILES CC(C)(C1=CC=C(C=C1)OC2=CC=C(C=C2)N3C(=O)C=CC3=O)C4=CC=C(C=C4)OC5=CC=C(C=C5)N6C(=O)C=CC6=O

Specifications

Melting Point 165°C
Color Yellow
Quantity 25 g
Formula Weight 570.60
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis[4-(4-maleimidophenoxy)phenyl]propane
EINECSNumber (5)-6344
TSCA No