accessibility menu, dialog, popup

UserName

CAS RN 79922-55-7

IUPAC Name: 1-{4-[4-(2-{4-[4-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)phenoxy]phenyl}propan-2-yl)phenoxy]phenyl}-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-{4-[4-(2-{4-[4-(2,5-dioxopyrrol-1-yl)phenoxy]phenyl}propan-2-yl)phenoxy]phenyl}pyrrole-2,5-dione
Molecular Weight (g/mol): 570.6
Molecular Formula: C35H26N2O6
InChi Key: XAZPKEBWNIUCKF-UHFFFAOYSA-N
SMILES: CC(C)(C1=CC=C(OC2=CC=C(C=C2)N2C(=O)C=CC2=O)C=C1)C1=CC=C(OC2=CC=C(C=C2)N2C(=O)C=CC2=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results