missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis[4-(4-aminophenoxy)phenyl]hexafluoropropane 98.0+%, TCI America™

$106.25 - $350.44

Chemical Identifiers

CAS 69563-88-8
Molecular Formula C27H20F6N2O2
Molecular Weight (g/mol) 518.46
MDL Number MFCD00015723
InChI Key HHLMWQDRYZAENA-UHFFFAOYSA-N
PubChem CID 94652
IUPAC Name 4-(4-{2-[4-(4-aminophenoxy)phenyl]-1,1,1,3,3,3-hexafluoropropan-2-yl}phenoxy)aniline
SMILES NC1=CC=C(OC2=CC=C(C=C2)C(C2=CC=C(OC3=CC=C(N)C=C3)C=C2)(C(F)(F)F)C(F)(F)F)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B13985G
View Documents
TCI America
B13985G
5 g
Each for $106.25
Only null left
 
B139825G
View Documents
TCI America
B139825G
25 g
Each for $350.44
Only null left
 
Description

Description

Specifications

Chemical Identifiers

69563-88-8
518.46
HHLMWQDRYZAENA-UHFFFAOYSA-N
4-(4-{2-[4-(4-aminophenoxy)phenyl]-1,1,1,3,3,3-hexafluoropropan-2-yl}phenoxy)aniline
C27H20F6N2O2
MFCD00015723
94652
NC1=CC=C(OC2=CC=C(C=C2)C(C2=CC=C(OC3=CC=C(N)C=C3)C=C2)(C(F)(F)F)C(F)(F)F)C=C1

Specifications

69563-88-8
White-Yellow
MFCD00015723
4,4′C-(Hexafluoroisopropylidene)bis[(4-aminophenoxy)benzene]
NC1=CC=C(OC2=CC=C(C=C2)C(C2=CC=C(OC3=CC=C(N)C=C3)C=C2)(C(F)(F)F)C(F)(F)F)C=C1
518.46
518.46
Crystalline Powder
163°C
C27H20F6N2O2
5 g
HHLMWQDRYZAENA-UHFFFAOYSA-N
4-(4-{2-[4-(4-aminophenoxy)phenyl]-1,1,1,3,3,3-hexafluoropropan-2-yl}phenoxy)aniline
94652
≥98.0% (HPLC,T)
2,2-Bis[4-(4-aminophenoxy)phenyl]hexafluoropropane
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents