accessibility menu, dialog, popup

UserName

CAS RN 69563-88-8

NH2OOH2NFFFFFF

CAS RN 69563-88-8

IUPAC Name: 4-(4-{2-[4-(4-aminophenoxy)phenyl]-1,1,1,3,3,3-hexafluoropropan-2-yl}phenoxy)aniline
Synonyms: 4-(4-{2-[4-(4-aminophenoxy)phenyl]-1,1,1,3,3,3-hexafluoropropan-2-yl}phenoxy)aniline
Molecular Weight (g/mol): 518.46
Molecular Formula: C27H20F6N2O2
InChi Key: HHLMWQDRYZAENA-UHFFFAOYSA-N
SMILES: NC1=CC=C(OC2=CC=C(C=C2)C(C2=CC=C(OC3=CC=C(N)C=C3)C=C2)(C(F)(F)F)C(F)(F)F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results