missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]propane 95.0+%, TCI America™

Catalog No. B2022500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2022500G 500 g
B202225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2022500G Supplier TCI America Supplier No. B2022500G

Chemical Identifiers

CAS 21850-44-2
Molecular Formula C21H20Br8O2
Molecular Weight (g/mol) 943.62
MDL Number MFCD00017887
InChI Key LXIZRZRTWSDLKK-UHFFFAOYNA-N
Synonym Tetrabromobisphenol A Bis(2,3-dibromopropyl) Ether
PubChem CID 62753
IUPAC Name 1,3-dibromo-5-{2-[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]propan-2-yl}-2-(2,3-dibromopropoxy)benzene
SMILES CC(C)(C1=CC(Br)=C(OCC(Br)CBr)C(Br)=C1)C1=CC(Br)=C(OCC(Br)CBr)C(Br)=C1

Specifications

Melting Point 117°C
Color White-Yellow
Quantity 500 g
Formula Weight 943.62
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]propane
EINECSNumber (4)-0212&(4)-0218
TSCA Yes