CAS RN 21850-44-2
IUPAC Name:
1,3-dibromo-5-{2-[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]propan-2-yl}-2-(2,3-dibromopropoxy)benzene
Synonyms:
1,3-dibromo-5-{2-[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]propan-2-yl}-2-(2,3-dibromopropoxy)benzene
Molecular Weight (g/mol):
943.62
Molecular Formula:
C21H20Br8O2
InChi Key:
LXIZRZRTWSDLKK-UHFFFAOYNA-N
SMILES:
CC(C)(C1=CC(Br)=C(OCC(Br)CBr)C(Br)=C1)C1=CC(Br)=C(OCC(Br)CBr)C(Br)=C1
1
–
1
of
1
results
1
2,2-Bis[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]propane 95.0+%, TCI America™
CAS: 21850-44-2 Molecular Formula: C21H20Br8O2 Molecular Weight (g/mol): 943.62 MDL Number: MFCD00017887 InChI Key: LXIZRZRTWSDLKK-UHFFFAOYNA-N Synonym: Tetrabromobisphenol A Bis(2,3-dibromopropyl) Ether PubChem CID: 62753 IUPAC Name: 1,3-dibromo-5-{2-[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]propan-2-yl}-2-(2,3-dibromopropoxy)benzene SMILES: CC(C)(C1=CC(Br)=C(OCC(Br)CBr)C(Br)=C1)C1=CC(Br)=C(OCC(Br)CBr)C(Br)=C1