missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Biphenyldicarboxylic Acid 98.0+%, TCI America™
$544.30 - $1621.73
Chemical Identifiers
| CAS | 482-05-3 |
|---|---|
| Molecular Formula | C14H10O4 |
| Molecular Weight (g/mol) | 242.23 |
| MDL Number | MFCD00002464 |
| InChI Key | GWZCCUDJHOGOSO-UHFFFAOYSA-N |
| Synonym | diphenic acid, 1,1'-biphenyl-2,2'-dicarboxylic acid, 2,2'-biphenyldicarboxylic acid, 2,2'-bibenzoic acid, biphenyl-2,2'-dicarboxylic acid, 2,2'-dicarboxybiphenyl, 2,2'-diphenic acid, 2-2-carboxyphenyl benzoic acid, o,o'-diphenic acid, o,o'-bibenzoic acid |
| PubChem CID | 10210 |
| ChEBI | CHEBI:23837 |
| IUPAC Name | 2-(2-carboxyphenyl)benzoic acid |
| SMILES | C1=CC=C(C(=C1)C2=CC=CC=C2C(=O)O)C(=O)O |
Chemical Identifiers
| 482-05-3 | |
| 242.23 | |
| GWZCCUDJHOGOSO-UHFFFAOYSA-N | |
| 10210 | |
| 2-(2-carboxyphenyl)benzoic acid |
| C14H10O4 | |
| MFCD00002464 | |
| diphenic acid, 1,1'-biphenyl-2,2'-dicarboxylic acid, 2,2'-biphenyldicarboxylic acid, 2,2'-bibenzoic acid, biphenyl-2,2'-dicarboxylic acid, 2,2'-dicarboxybiphenyl, 2,2'-diphenic acid, 2-2-carboxyphenyl benzoic acid, o,o'-diphenic acid, o,o'-bibenzoic acid | |
| CHEBI:23837 | |
| C1=CC=C(C(=C1)C2=CC=CC=C2C(=O)O)C(=O)O |
Specifications
| 482-05-3 | |
| White | |
| MFCD00002464 | |
| diphenic acid, 1,1'-biphenyl-2,2'-dicarboxylic acid, 2,2'-biphenyldicarboxylic acid, 2,2'-bibenzoic acid, biphenyl-2,2'-dicarboxylic acid, 2,2'-dicarboxybiphenyl, 2,2'-diphenic acid, 2-2-carboxyphenyl benzoic acid, o,o'-diphenic acid, o,o'-bibenzoic acid | |
| C1=CC=C(C(=C1)C2=CC=CC=C2C(=O)O)C(=O)O | |
| 242.23 | |
| CHEBI:23837 | |
| ≥98.0% (GC,T) | |
| 2,2′-Biphenyldicarboxylic Acid |
| 233°C | |
| C14H10O4 | |
| 25 g | |
| GWZCCUDJHOGOSO-UHFFFAOYSA-N | |
| 2-(2-carboxyphenyl)benzoic acid | |
| 10210 | |
| 242.23 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-0083
TSCA : Yes