accessibility menu, dialog, popup

UserName

CAS RN 482-05-3

OHOHOO

CAS RN 482-05-3

IUPAC Name: [1,1'-biphenyl]-2,2'-dicarboxylic acid
Synonyms: diphenic acid O,O'-bibenzoic acid diphenic acid (8CI) 2,2'-diphenic acid O,O'-diphenic acid
Molecular Weight (g/mol): 242.23
Molecular Formula: C14H10O4
InChi Key: GWZCCUDJHOGOSO-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1C1=CC=CC=C1C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results