missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Biphenyldicarboxylic Acid 98.0+%, TCI America™

Catalog No. D086425G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D086425G 25 g
D0864100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D086425G Supplier TCI America Supplier No. D086425G
Only null left

Chemical Identifiers

CAS 482-05-3
Molecular Formula C14H10O4
Molecular Weight (g/mol) 242.23
MDL Number MFCD00002464
InChI Key GWZCCUDJHOGOSO-UHFFFAOYSA-N
Synonym diphenic acid, 1,1'-biphenyl-2,2'-dicarboxylic acid, 2,2'-biphenyldicarboxylic acid, 2,2'-bibenzoic acid, biphenyl-2,2'-dicarboxylic acid, 2,2'-dicarboxybiphenyl, 2,2'-diphenic acid, 2-2-carboxyphenyl benzoic acid, o,o'-diphenic acid, o,o'-bibenzoic acid
PubChem CID 10210
ChEBI CHEBI:23837
IUPAC Name 2-(2-carboxyphenyl)benzoic acid
SMILES C1=CC=C(C(=C1)C2=CC=CC=C2C(=O)O)C(=O)O

Specifications

Melting Point 233°C
Color White
Quantity 25 g
Formula Weight 242.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Biphenyldicarboxylic Acid
EINECSNumber (4)-0083
TSCA Yes