missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2',7,7'-Tetrabromo-9,9'-spirobi[9H-fluorene] 98.0+%, TCI America™
$113.71 - $382.13
Chemical Identifiers
| CAS | 128055-74-3 |
|---|---|
| Molecular Formula | C25H12Br4 |
| Molecular Weight (g/mol) | 631.99 |
| MDL Number | MFCD08704220 |
| InChI Key | MASXXNUEJVMYML-UHFFFAOYSA-N |
| PubChem CID | 371282 |
| IUPAC Name | 2,2',7,7'-tetrabromo-9,9'-spirobi[fluorene] |
| SMILES | BrC1=CC2=C(C=C1)C1=C(C=C(Br)C=C1)C21C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2 |
Chemical Identifiers
| 128055-74-3 | |
| 631.99 | |
| MASXXNUEJVMYML-UHFFFAOYSA-N | |
| 2,2',7,7'-tetrabromo-9,9'-spirobi[fluorene] |
| C25H12Br4 | |
| MFCD08704220 | |
| 371282 | |
| BrC1=CC2=C(C=C1)C1=C(C=C(Br)C=C1)C21C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2 |
Specifications
| 128055-74-3 | |
| C25H12Br4 | |
| 1 g | |
| BrC1=CC2=C(C=C1)C1=C(C=C(Br)C=C1)C21C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2 | |
| 631.99 | |
| 631.99 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD08704220 | |
| MASXXNUEJVMYML-UHFFFAOYSA-N | |
| 2,2',7,7'-tetrabromo-9,9'-spirobi[fluorene] | |
| 371282 | |
| ≥98.0% (T) | |
| 2,2′,7,7′-Tetrabromo-9,9′-spirobi[9H-fluorene] |
Safety and Handling
TSCA : No