missing translation for 'onlineSavingsMsg'
Learn More

2,2':6',2″-Terpyridine-4'-carboxylic Acid 98.0+%, TCI America™

Supplier:  TCI America T32451G

 View more versions of this product

Catalog No. T32451G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,2′:6′,2′′-Terpyridine-4′-carboxylic Acid
290°C
C16H11N3O2
1 g
ZYTWXMBGOUJDHJ-UHFFFAOYSA-N
6-(pyridin-2-yl)-[2,2'-bipyridine]-4-carboxylic acid
2762749
≥98.0% (GC,T)
148332-36-9
White-Yellow
MFCD04114290
2,2':6',2-terpyridine-4'-carboxylic acid, 2,2':6',2-terpyridine-4'-carboxylicacid, 2,6-bis pyridin-2-yl pyridine-4-carboxylic acid, 6-pyridin-2-yl-2,2'-bipyridine-4-carboxylic acid, acmc-1c3v7, 4'-carboxy-2,2':6',2-terpyridine, 2,2:6,2'-terpyridine-4-carboxylic acid, 1∼2∼,2∼2∼:2∼6∼,3∼2∼-terpyridine-2∼4∼-carboxylic acid, 2,2\\':6\\',2\\'\\'-terpyridine-4\\'-carboxylic acid
OC(=O)C1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1
277.28
277.28
Crystalline Powder

Chemical Identifiers

148332-36-9
277.28
ZYTWXMBGOUJDHJ-UHFFFAOYSA-N
2762749
OC(=O)C1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1
C16H11N3O2
MFCD04114290
2,2':6',2-terpyridine-4'-carboxylic acid, 2,2':6',2-terpyridine-4'-carboxylicacid, 2,6-bis pyridin-2-yl pyridine-4-carboxylic acid, 6-pyridin-2-yl-2,2'-bipyridine-4-carboxylic acid, acmc-1c3v7, 4'-carboxy-2,2':6',2-terpyridine, 2,2:6,2'-terpyridine-4-carboxylic acid, 1∼2∼,2∼2∼:2∼6∼,3∼2∼-terpyridine-2∼4∼-carboxylic acid, 2,2\\':6\\',2\\'\\'-terpyridine-4\\'-carboxylic acid
6-(pyridin-2-yl)-[2,2'-bipyridine]-4-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions