missing translation for 'onlineSavingsMsg'
Learn More

2,2':6',2″-Terpyridine-4'-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7137654
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3245200MG 200 mg
T32451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3245200MG Supplier TCI America Supplier No. T3245200MG
Only null left

Chemical Identifiers

CAS 148332-36-9
Molecular Formula C16H11N3O2
Molecular Weight (g/mol) 277.28
MDL Number MFCD04114290
InChI Key ZYTWXMBGOUJDHJ-UHFFFAOYSA-N
Synonym 2,2':6',2-terpyridine-4'-carboxylic acid, 2,2':6',2-terpyridine-4'-carboxylicacid, 2,6-bis pyridin-2-yl pyridine-4-carboxylic acid, 6-pyridin-2-yl-2,2'-bipyridine-4-carboxylic acid, acmc-1c3v7, 4'-carboxy-2,2':6',2-terpyridine, 2,2:6,2'-terpyridine-4-carboxylic acid, 1∼2∼,2∼2∼:2∼6∼,3∼2∼-terpyridine-2∼4∼-carboxylic acid, 2,2\\':6\\',2\\'\\'-terpyridine-4\\'-carboxylic acid
PubChem CID 2762749
IUPAC Name 6-(pyridin-2-yl)-[2,2'-bipyridine]-4-carboxylic acid
SMILES OC(=O)C1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1

Specifications

Melting Point 290°C
Color White-Yellow
Quantity 200 mg
Formula Weight 277.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′:6′,2′′-Terpyridine-4′-carboxylic Acid