accessibility menu, dialog, popup

UserName

NNNOHO

CAS RN 148332-36-9

IUPAC Name: 6-(pyridin-2-yl)-[2,2'-bipyridine]-4-carboxylic acid
Synonyms: 6-(pyridin-2-yl)-[2,2'-bipyridine]-4-carboxylic acid
Molecular Weight (g/mol): 277.28
Molecular Formula: C16H11N3O2
InChi Key: ZYTWXMBGOUJDHJ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)