missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2',5,5'-Tetramethylbiphenyl, 98%
Description
2,2',5,5'-Tetramethylbiphenyl is used to make other chemicals and as an intermediate in pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,2′,5,5′-Tetramethylbiphenyl is used to make other chemicals and as an intermediate in pharmaceuticals.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
2,2′,5,5′-Tetramethylbiphenyl is used to make other chemicals and as an intermediate in pharmaceuticals.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | 48°C to 51°C |
| Quantity | 1 g |
| Beilstein | 2046200 |
| Synonym | 2,2',5,5'-tetramethylbiphenyl, 2,2',5,5'-tetramethyl-1,1'-biphenyl, 1,1'-biphenyl, 2,2',5,5'-tetramethyl, biphenyl-, 2,2',5,5'-tetramethyl, 2-2,5-dimethylphenyl-1,4-dimethylbenzene, acmc-20aokf, 2,5,2',5'-tetramethylbiphenyl, 1,1'-biphenyl,2,2',5,5'-tetramethyl, 2,2',5,5'-tetramethyl-1,1'-biphenyl # |
| Boiling Point | 284°C (732 mmHg) |
| Solubility Information | Insoluble in water. |
| Formula Weight | 210.32 |
| Percent Purity | 98% |
| Chemical Name or Material | 2,2',5,5'-Tetramethylbiphenyl |
Safety and Handling
| EINECSNumber | 000-000-0 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only